Learn More
Thermo Scientific™ GC Installation Kit
Catalog No. 60180888 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Gas Chromatography Systems
Catalog No. For Use With (Equipment)
60-180-888 Gas Chromatography Systems
1 options
Catalog No. 60-180-888 Supplier Thermo Scientific™ Supplier No. 60180888
Add to Cart
Add to Cart
Add a GC system to your lab bench with the Thermo Scientific™ GC Installation Kit. The required tubing and fittings are conveniently supplied in this kit.
Add a GC system to your lab bench with the Thermo Scientific™ GC Installation Kit. The required tubing and fittings are conveniently supplied in this kit.
Includes:
Tubing cutter; 1/8 x 1/4in. reamer; 7/16in. wrench; 1/2in. wrench; 1/8in. brass tees, 4; 1/8in. brass nuts, 10; Brass front and back ferrules, 10; 15.2m instrument-grade, cleaned 1/8in. copper tubing
TRUSTED_SUSTAINABILITY
Specifications
Type GC Installation Kit
Description GC Installation Kit
For Use With (Application) GC
Quantity Each
For Use With (Equipment) Gas Chromatography Systems
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.