Learn More
Thermo Scientific™ GC Tool Kit for Agilent Instruments
Catalog No. 60180786 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Agilent GC-MS Instruments
Catalog No. For Use With (Equipment)
60-180-786 Agilent GC-MS Instruments
1 options
Catalog No. 60-180-786 Supplier Thermo Scientific™ Supplier No. 60180786
Add to Cart
Add to Cart
Thermo Scientific™ GC Tool Kit for Agilent Instruments includes common consumable supplies and tools.
Thermo Scientific™ GC Tool Kit for Agilent Instruments includes common consumable supplies and tools.
  • Replacements included perform like original instrument parts
  • Parts list included for easy reordering
Includes:
Capillary installation gauge, injector wrench, septum nut removal tool, open-end wrenches in 7/16 x 1/2in. and 1/2 x 9/16in. sizes.
TRUSTED_SUSTAINABILITY
Specifications
Type Capillary Tool Kit
Description GC Tool Kit for Agilent Instruments
For Use With (Application) GC
For Use With (Equipment) Agilent GC-MS Instruments
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.