Learn More
Invitrogen™ GCAT Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RGAGTWKSER VITSRQGPHI RVDGVSGGIL NFCANNYLGL SSHPEVIQAG LQALEEFGAG LSSVRFICGT Q Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 93%.
Specifications
Specifications
| Antigen | GCAT |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Gcat |
| Gene Accession No. | O75600 |
| Gene Alias | 2-amino-3-ketobutyrate coenzyme A ligase; 2-amino-3-ketobutyrate coenzyme A ligase, mitochondrial; 2-amino-3-ketobutyrate-CoA ligase; AI526977; AKB ligase; aminoacetone synthase; aminoacetone synthetase; aminotransferase; glycine acetyltransferase; aminoacetone synthetase; KBL; GCAT; Glycine acetyltransferase; glycine C-acetyltransferase; glycine C-acetyltransferase (2-amino-3-ketobutyrate coenzyme A ligase); glycine C-acetyltransferase (2-amino-3-ketobutyrate-coenzyme A ligase); KBL; Unknown (protein for MGC:139551) |
| Gene Symbols | Gcat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.