Learn More
AdipoGen Glycochenodeoxycholic acid sodium salt (500 mg)
SDP
Supplier:  AdipoGen CDXG0033M500
Encompass_Preferred
Glycochenodeoxycholic acid sodium salt (500 mg), AdipoGen Life Sciences. Chemical. CAS: 16564-43-5. Formula: C26H42NNaO5. MW: 471.61. Synthetic. Glycochenodeoxycholic acid (GCDCA) is a bile salt formed in the liver that acts as a biosurfactant to solubilize lipids for absorption and is itself absorbed. GCDCA functions as a choleretic, increasing the volume of bile secreted from the liver, and a cholagogue, promoting the discharge of bile from the digestive system. GCDCA has been used to demonstrate a role for bile acids in promoting colorectal carcinogenesis and to inhibit calcium hydroxyapatite precipitation to study the pathogenesis of black pigment gallstones. This product has also been shown to induce apoptosis in isolated hepatocytes via modulation of PKC activity. This compound is used as an anionic detergent to solubilize lipids and as a chiral reagent.
Catalog No. 50-149-0418
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.