Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ GCDFP-15 (Gross Cystic Disease Fluid Protein-15) Ab-1, Mouse Monoclonal Antibody

Catalog No. p-4545761 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1170S0 100 μL Concentrated
MS1170S1 500 μL Concentrated
MS1170S 1 mL Concentrated
MS1170R7 7 mL Prediluted
MS1170RQ 12 mL Ready-to-Use
MS1170R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS1170S0 Supplier Epredia™ Supplier No. MS1170S0
Only null left
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry experiments.

Gross cystic disease of the breast is benign premenopausal disorder in which cysts are a predominant pathological lesion. These cysts appear to be formed from excessive apocrine cystic secretions. This fluid is composed of several glycoproteins including a unique 15 kDa monomer protein, Gross Cystic Disease Fluid Protein-15 (GCDFP15). Cytosolic analysis of normal tissue specimens from all major organs has demonstrated GCDFP15 in apocrine epithelia, lacrimal, ceruminous and Moll's glands and in numerous serous cells of the submandibular, tracheal, bronchial, sublingual and minor salivary glands. GCDFP15 and prostate specific antigen are co-expressed in androgen receptor-positive breast tumours.

Host Species: Mouse

Clone: 23A3

Isotype: IgG2a/κ

Species Reactivity: Human and Rat. Others not known.

Immunogen: Recombinant protein encoding the excreted domain of human GCDFP-15

Molecular Weight: 15 kDa

Positive Control: Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen GCDFP-15 (Gross Cystic Disease Fluid Protein-15) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 23A3
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding the excreted domain of human GCDFP15
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.