Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeneBLAzer™ GeneBLAzer™ GCGR-CRE-bla CHO-K1 Cells

Catalog No. K1855
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1855 1 vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1855 Supplier GeneBLAzer™ Supplier No. K1855
Add to Cart
Add to Cart
Request a Quote

GeneBLAzer™ GCGR-CRE-bla CHO-K1 Cells contain the human glucagon receptor (GCGR).

  • Accession Number: NM_000160
  • Population Status: Pool of Stable Cells
  • GeneBLAzer™ GCGR-CRE-bla CHO-K1 cells contain the human glucagon receptor (GCGR), (Accession # NM_000160) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 (Cat No. K1535) cells contain a beta-lactamase reporter gene under control of the CRE Response Element. The GCGR-CRE-bla CHO-K1 cells are functionally validated for Z’-factor and EC50 concentrations of Glucagon. In addition, GCGR-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000160
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage GeneBLAzer™ GCGR-CRE-bla CHO-K1 cells are shipped on dry ice, 8 x 10^6 cells⁄vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Detection Method Fluorescence
For Use With (Application) FRET
Human Protein Class G-Protein Coupled Receptor (GPCR)
Population Status Pool of Stable Cells
Product Line GeneBLAzer
Quantity 1 vial
Readout End Point
Target Entry GCGR
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.