Learn More
GCKR Mouse anti-Human, Clone: OTI1E7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502152

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein belonging to the GCKR subfamily of the SIS (Sugar ISomerase) family of proteins. The gene product is a regulatory protein that inhibits glucokinase in liver and pancreatic islet cells by binding non-covalently to form an inactive complex with the enzyme. This gene is considered a susceptibility gene candidate for a form of maturity-onset diabetes of the young (MODY).Specifications
GCKR | |
Monoclonal | |
Unconjugated | |
GCKR | |
FGQTL5, GKRP | |
Mouse | |
Affinity Chromatography | |
RUO | |
2646 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1E7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q14397 | |
GCKR | |
Full length human recombit protein of human GCKR produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.