Learn More
Invitrogen™ GCN2 Polyclonal Antibody

Description
GCN2 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | GCN2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | EIF2AK4 |
| Gene Accession No. | Q9P2K8 |
| Gene Alias | 2610011M03; EIF2AK4; eIF-2-alpha kinase GCN2; eukaryotic translation initiation factor 2 alpha kinase 4; eukaryotic translation initiation factor 2-alpha kinase 4; Gcn2; GCN2 eIF2alpha kinase; GCN2-like protein; general control nonderepressible 2; Kiaa1338; mGCN2; PVOD2 |
| Gene Symbols | EIF2AK4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.