Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCN5 Polyclonal Antibody, Active Motif™
SDP

Catalog No. p-7188666
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4125 10 μL
50-199-4124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-4125 Supplier Active Motif Supplier No. 39832
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SAGA (Spt-Ada-Gen5 histone acetyltransferase complex) is a histoneacetylase complex which has Gcn5p as catalyst subunit and functions overlapping with the fundamental transcription factor TFIID which has Taf1p as catalyst subunit. However, SAGA and TFIID have different allotment and each accomplishes the important role in the transcription for Housekeeping gene group and Stress Responding gene group. Also, SAGA is a gigantic protein complex which is composed of Ada protein group (5 kinds), TBP related protein group (4 kinds), TAF protein group (5 kinds that also pertain to TFIID), and other protein groups (>6 kinds). As its molecular function, there is the chemical modification of histone, or the recruitment by direct interaction of transcription regulating factor on DNA, or the control of transcription starting reaction by TBP. Gcn5p that show histoneacetylase activity is one kind of the above-mentioned Ada protein group, and in the case of budding yeast it is composed of 439 amino acid residues.

Specifications

Antigen GCN5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene KAT2A
Gene Accession No. Q92830
Gene Alias 1110051E14Rik; AW212720; GCN5; GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 2; GCN5 general control of amino acid synthesis-like 2; GCN5L2; general control of amino acid synthesis 5-like 2; General control of amino acid synthesis protein 5-like 2; general control of amino acid synthesis, yeast homolog-like 2; General control of amino acid synthesis, yeast, homolog-like 2; general control of amino acid synthesis-like 2; hGCN5; Histone acetyltransferase GCN5; Histone acetyltransferase KAT2A; Histone succinyltransferase KAT2A; hsGCN5; K(lysine) acetyltransferase 2A; KAT2A; KATA2; Lysine acetyltransferase 2A; MmGCN5; PCAF-b; PCAF-B/GCN5; STAF97
Gene Symbols KAT2A
Host Species Rabbit
Immunogen This GCN5 antibody was raised against a peptide within the N-terminal region of human GCN5.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2648
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.