Learn More
Invitrogen™ GCNF Monoclonal Antibody (4G8)

Description
MA5-17140 targets NR6A1 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human samples. The MA5-17140 immunogen is purified recombinant fragment of human NR6A1 (amino acids: 65-118) expressed in E. Coli. MA5-17140 detects NR6A1 which has a predicted molecular weight of approximately 54.3kDa.
Specifications
Specifications
| Antigen | GCNF |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4G8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | NR6A1 |
| Gene Accession No. | Q15406 |
| Gene Alias | 1700113M01Rik; CT150; Gcnf; GCNF1; Germ cell nuclear factor; germ cell nuclear factor variant 1; h hRTR; hGCNF; hRTR; mGCNF; NCNF; NR61; NR6A1; Nuclear receptor subfamily 6 group A member 1; nuclear receptor subfamily 6, group A, member 1; retinoic acid receptor-related testis-associated receptor; retinoid receptor-related testis-specific receptor; RTR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.