Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583495 Supplier Invitrogen™ Supplier No. PA583495
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLYTLFLESV QSRLRHGEDS TPQVLTEQQA TKENLMKMQS IAESHLELDD VHDPLLAINF ARMYLEQSDF HEKFAGGDVC VDRSSESVTC QTFELTLRSC Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 85%.

The gamma-Tubulin complex is composed of gamma Tubulin and the gamma-Tubulin complex associated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. gamma-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the gamma-Tubulin complex. GCP5 ( gamma-Tubulin complex component 5), also known as TUBGCP5, is a 1, 024 amino acid member of the gamma-Tubulin complex and is highly expressed in heart and skeletal muscle. Defects in the gene encoding GCP5 are implicated in Prader-Willi syndrome (PWS), a rare genetic disorder associated with obesity, compulsive behavior and lower intellectual ability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCP5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tubgcp5
Gene Accession No. Q96RT8
Gene Alias B130010C12Rik; Gamma-tubulin complex component 5; gamma-tubulin complex component GCP5; GCP5; GCP-5; KIAA1899; mKIAA1899; Tubgcp5; tubulin gamma complex associated protein 5; tubulin, gamma complex associated protein 5
Gene Symbols Tubgcp5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TUBGCP5. Recombinant protein control fragment (Product #RP-105983).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114791
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.