Learn More
ACROBiosystems ACROBiosystems Human GDF-15 / MIC-1 Protein, His Tag
Supplier:  ACROBiosystems GD5H51491MG
Encompass_Preferred
Recombinant Protein;1MG;Human GDF-15, His Tag (GD5-H5149) is expressed from E.coli cells. It contains AA Ala 197 - Ile 308 (Accession # Q99988-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 14.2 kDa. As a result of glycosylation, the protein migrates as 16-17 kDa under reducing (R) condition, and 27 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human GDF-15, His Tag (Cat. No. GD5-H5149) at 1 μg/mL (100 μL/well) can bind Human GFR alpha-like Fc Chimera Protein with a linear range of 0.2-8 ng/mL (QC tested).
Catalog No. 50-195-6517
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.