Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ GDF-8/Myostatin DuoSet ELISA
SDP

Catalog No. DY78805 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit (5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY78805 1 Kit (5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY78805 Supplier R&D Systems™ Supplier No. DY78805
Add to Cart
Add to Cart
This item is not returnable. View return policy

GDF-8/Myostatin DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The GDF-8/Myostatin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies multi-species GDF-8/Myostatin.

Specifications

Assay Range 31.20 - 2000pg/mL
Target Species Multi-species
For Use With (Application) ELISA
Gene ID (Entrez) 2660
Quantity 1 Kit (5 Plates)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.