Learn More
Genscript Corporation GDF15 hFc ChimeraBiotin Cyn
Supplier:  Genscript Corporation Z04366100
Encompass_Preferred
Growth and differentiation factor 15 (GDF15) is an inflammation-associated hormone with poorly defined biology Here we investigated the role of GDF15 in bacterial and viral infections Inflammation induced GDF15 and that GDF15 was necessary for surviving both bacterial and viral infections as well as sepsis The protective effects of GDF15 were largely independent of pathogen control or the magnitude of inflammatory response suggesting a role in disease tolerance
Catalog No. 50-000-58712
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.