Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GDF15 (Human) ELISA Kit

Catalog No. 89101038 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
96 Assays
Sample Type:
CCS, Plasma (EDTA, Heparin), Serum
CCS, Plasma, Serum, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sample Type
89-101-038 1 Kit CCS, Plasma (EDTA, Heparin), Serum
89-028-582 96 Assays CCS, Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-038 Supplier Abnova™ Supplier No. KA3359
Add to Cart
Add to Cart

In-vitro ELISA for the quantitative measurement of Human GDF15

Specifications

Calibration Range 31.2 to 2000pg/mL
Cross Reactivity Human
Description GDF15 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human GDF15.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type CCS, Plasma (EDTA, Heparin), Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type GDF15 (Human) ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.