Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GDF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547953
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547953 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547953 Supplier Invitrogen™ Supplier No. PA547953
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse (rm) GDF-6 is observed, and less than 5% cross-reactivity with rmGDF-1, rmGDF-3, rmGDF-8, rmGDF-9, recombinant human GDF-11, and rhGDF-15 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

GDF-5 is expressed in long bones during embryonic development and postnatally in articular cartilage. Mutations in the GDF-5 gene have been implicated in Hunter-Thompson type dwarfism and in Grebe Syndrome, which is characterized by short stature, extra digits, and short and deformed extremities. The mature and functional form of GDF-5 is a homodimer of two 120 amino-acid polypeptide chain (monomers) linked by a single disulfide bond. Each GDF-5 monomer is expressed as the C-terminal part of a precursor polypeptide, which also contains a 27 amino acid signal peptide and a 354 amino acid propeptide. This precursor undergoes intracellular dimerization, and upon secretion it is processed by a furin-type protease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF5
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene GDF5
Gene Accession No. P43027
Gene Alias BDA1C; Bmp14; BMP-14; Bone morphogenetic protein 14; bp; brachypodism; brp; Cartilage-derived morphogenetic protein 1; cartilage-derived morphogenetic protein-1; CDMP1; Cdmp-1; GDF5; GDF-5; growth differentiation factor 5; growth/differentiation factor 5; H-GDF-5; LAP4; LAP-4; Lipopolysaccharide-associated protein 4; LPS-associated protein 4; OS5; radotermin; SYM1B; SYNS2
Gene Symbols GDF5
Host Species Goat
Immunogen E. coli-derived recombinant mouse GDF-5 Ala376-Arg495.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14563
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.