Learn More
Invitrogen™ GDF9 Polyclonal Antibody

Description
GDF9 Polyclonal Antibody detects endogenous levels of GDF9 protein.
Specifications
Specifications
| Antigen | GDF9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | GDF9 |
| Gene Accession No. | O60383, Q07105 |
| Gene Alias | Gdf9; Gdf-9; growth differentiation factor 9; growth/differentiation factor 9 |
| Gene Symbols | GDF9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.