Learn More
Invitrogen™ GDNF Polyclonal Antibody

Description
PA1-9524 detects GDNF in human, mouse and rat samples. PA1-9524 has been successfully used in Western blot, ELISA, and Immunohistochemistry procedures. By Western blot, PA1-9524 detects a 24 kDa protein representing GDNF. PA1-9524 immunogen is a synthetic peptide based on the human GDNF protein. Store product at 4°C for up to one week, and -20°C for long term storage.
Specifications
Specifications
| Antigen | GDNF |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | GDNF |
| Gene Accession No. | P39905, P48540, Q07731 |
| Gene Alias | AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor |
| Gene Symbols | GDNF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.