Learn More
AdipoGen GE81112 A B
SDP
Supplier:  AdipoGen AGCN20306M001
Encompass_Preferred
Chemical. CAS 883726-13-4 GE81112A, 883726-14-5 GE81112B. Formula C24H34ClN9O10 A, C24H35ClN10O10 B. MW 644.0 A659.1 B. Isolated from Strepomyces sp. Tetrapeptide antibiotic. Potent and selective inhibitor of bacterial protein synthesis. Translational inhibitor specific for the initiation phase. Binds to the 30S ribosomal subunit and specifically inhibits P-site decoding of the mRNA initiation codon by the fMet-tRNA anticodon. Inhibits in vivo protein synthesis but not other cell functions such as DNA duplication, transcription and cell wall synthesis. Antibacterial activity against some Gram-positive and Gram-negative bacteria. Unique scaffold for designing new drugs.
Catalog No. 50-256-2624
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.