Learn More
Medchemexpress LLC HY-50895A 5g , Gefitinib (hydrochloride) ZD-1839 hydrochloride CAS:184475-55-6 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY50895A5G
Encompass_Preferred
Medchemexpress, HY-50895A 5g Gefitinib (hydrochloride) ZD-1839 hydrochloride CAS:184475-55-6 Formula:C22H25Cl2FN4O3 IC50: 37 nM (Tyr1173 site, in NR6wtEGFR cells), 37 nM (Tyr992 site, in NR6wtEGFR cells) Purity:98% Gefitinib hydrochloride is an inhibitor that specifically binds and inhibits the EGFR tyrosine kinase , with the IC 50 value of 2-37 nM in NR6wtEGFR cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2236
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.