Learn More
Zymo Research Corporation Sample Zymoclean™ Gel DNA Recovery Kit (10 Preps) w/ Zymo-Spin™ I Columns (Uncapped)
SDP
Supplier:  Zymo Research Corporation D4001S
Encompass
Free Sample Simply add the specially formulated Agarose Dissolving Buffer (ADB) to the gel slice containing your DNA sample, let dissolve, and then transfer to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product utilizes Fast-Spin column technology to yield high-quality DNA in just 15 minutes. DNA purified using the Zymoclean™ Gel DNA Recovery Kit is perfectly suited for use in DNA ligation reactions, sequencing, DNA labeling reactions, PCR, etc.
Catalog No. NC3345980
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.