Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Gel Filtration Calibration Kits
GSA_VA

Catalog No. 45001433 Shop All Cytiva Products
Change view
Click to view available options
MWCO:
43,000 to 669,000
6500 to 75,000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO
45-001-433 43,000 to 669,000
45-001-432 6500 to 75,000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-001-433 Supplier Cytiva Supplier No. 28403842
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Calibrate gel filtration columns simply, reliably. Cytiva Life Sciences™ Gel Filtration Calibration Kits contain a well-defined series of protein standards.

  • Optimized range of proteins suited to high-resolution media and prepacked columns
  • Each kit contains five vials of lyophilized proteins
  • Blue Dextran 2000 included to determine the void fraction of the column

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity Each
For Use With (Equipment) High-resolution media and prepacked column
Product Type Calibration Kit
For Use With (Application) Calibration of high-resolution gel filtration media and column such as Superdex™, Superose and Sephacryl
Includes 50mg Ovalbumin (43,000 Mr), 50mg Conalbumin (75,000 Mr), 50mg Aldolase (158,000 Mr), 15mg Ferritin (440,000 Mr), 50mg Thyroglobulin (669,000 Mr), 50mg Blue Dextran (2,000,000 Mr)
MWCO 43,000 to 669,000
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.