Learn More
Electron Microscopy Sciences EMS SmartDoc™ Gel Imaging Enclosure
SDP
Supplier:  Electron Microscopy Sciences 8705701
Encompass_Preferred
The SmartDoc system uses a smart phone or tablet to quickly obtain quality images of gels for your records or anaylsis. With an adjustable focal distance for optimum focus, the SmartDoc enclosure is simply place over your gel and transilluminator. Lay your smart phone or tablet onto the non-slip, rubber pad, and align the camera lens over the filter. Raising and lowering the platform allows optimization of focus and image size.
Filters are available for compatibility with multiple light sources and dyes. The Blue Light model uses high intensity blue LEDs to excite green dyes and this eliminates the need for hazardous and DNA-damaging UV light.
Images can be previewed on the device's display before capturing the picture. Images are easily shared, printed or transferred via text message, email or cloud sharing apps. Gel analysis software can be used to analyze the image. Simply place the enclosure over your gel and transilluminator.
Catalog No. 50-192-7930
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.