Learn More
Med Vet International Ultrasound Gel Aquasonic Clear Transmission 8.5oz Bottle, 03-08, Each
Supplier: Med Vet International 227100EA
No fragrance or color For diagnostic and therapeutic medical ultrasound procedures Acoustically correct for the broad range of frequencies used Hypoallergenic, bacteriostatic, nonirritating Water soluble, No formaldehyde, Not a spermicide No fragrance or color 5 liter SONICPAC with refillable dispenser