Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Gelatin Blocking Buffer, 1% in PBS

Catalog No. AAJ62755K2
Click to view available options
Quantity:
500 mL
1 L

Catalog No. AAJ62755K2

Supplier: Thermo Scientific Chemicals J62755K2

Only null left
Add to Cart

Suitable for use as a non-specific blocking agent for Western blots and ELISA assays. It is used to block non-specific binding on the membrane, when using nylon or nitrocellulose (but not PVDF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Blocking Buffer
Concentration 1% in PBS
Physical Form Liquid
Quantity 1 L

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.