Learn More
Sigma Aldrich Fine Chemicals Biosciences Gelatin from bovine skin, 100G, 9000-70-8, MFCD00081638
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G9382100G
Encompass
Gelatin from bovine skin, 100G. gel strength ~225 g Bloom, Type B
Catalog No. NC0700810
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.