Learn More
Electron Microscopy Sciences Gelatin, Powder, U.S.P., Type A, 300 Bloom, 9000-70-8, 100G
SDP
Supplier:  Electron Microscopy Sciences 16564
Encompass_Preferred
Solubility: 12% in H2O at 20°C, pH (1% in H2O): 3.5-5.0; Gelatin Powder is used as a combination Acrylamide-Gelatin-Jung resin for cryosectioning of large and soft tissues (eggs, fishes, insects). A new embedding medium for cryosectioning eggs of high yolk and lipid content. It is used to prepare gelatin-chrome alum adhesive to coat glass slides for mounting semi-thin sections for staining.
Catalog No. 50-260-87
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.