Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Gelatin Sepharose™ 4B Media

Catalog No. 45000170 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-170 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-170 Supplier Cytiva Supplier No. 17095601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for the purification or depletion of fibronectin as gelatin binds to fibronectin with high specificit

  • Bovine gelatin coupled to Sepharose 4B, in a 4% agarose matrix
  • Ligand coupling method: cyanogen bromide activation
  • Ligand concentration: 4.5-8mg gelatin/mL medium
  • Average particle size: 90μm
  • Stable in the pH range 3 to 10, and in the presence of all commonly used buffers

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
Material Gelatin Sepharose 4 B
Particle Size 90 μm
Matrix Affinity Resin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.