Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gelatin, type A, 175 Bloom

Catalog No. AAJ6154822
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6154822 100 g
AAJ6154836 500 g
AAJ61548A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6154822 Supplier Thermo Scientific Chemicals Supplier No. J6154822
Add to Cart

CAS: 9000-70-8

Gelatin is used to make lithographic and printing inks, plastic compounds, artificial silk, matches and pharmaceutical capsules. It finds application in light filters for mercury lamps, clarifying agent, pharmaceutical aid (suspending agent) and gelling component. It acts as an additive for foods and microencapsulating agent. Further, it is employed in electroplating, hectographic masters, paper and textile sizing, cosmetics, ointments and other medical and veterinary applications. It is useful in histochemistry, bacterial culture media and light microscope autoradiography. In addition to this, it is used to make the shells of paintballs and to hold silver halide crystals in an emulsion, in virtually all photographic films and photographic papers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gelatin is used to make lithographic and printing inks, plastic compounds, artificial silk, matches and pharmaceutical capsules. It finds application in light filters for mercury lamps, clarifying agent, pharmaceutical aid (suspending agent) and gelling component. It acts as an additive for foods and microencapsulating agent. Further, it is employed in electroplating, hectographic masters, paper and textile sizing, cosmetics, ointments and other medical and veterinary applications. It is useful in histochemistry, bacterial culture media and light microscope autoradiography. In addition to this, it is used to make the shells of paintballs and to hold silver halide crystals in an emulsion, in virtually all photographic films and photographic papers.

Solubility
Soluble in water, glycerol and acetic acid.

Notes
Of porcine originIncompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-70-8
MDL Number MFCD00081638

Specifications

Color White to Yellow
Quantity 100 g
Merck Index 14,4382
Solubility Information Soluble in water,glycerol and acetic acid.
Physical Form Powder
Chemical Name or Material Gelatin, type A, 175 Bloom
Hazard Category H500
EINECSNumber 232-554-6
RTECSNumber LX8580000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.