Learn More
Selleck Chemical LLC Geldanamycin 5mg 30562-34-6 NSC 122750
SDP
Supplier:  Selleck Chemical LLC S27135MG
Encompass_Preferred
Geldanamycin (NSC 122750) is a natural existing HSP90 inhibitor with Kd of 1.2 ?M, specifically disrupts glucocorticoid receptor (GR)/HSP association. Geldanamycin attenuates virus infection-induced ALI (acute lung injury)/ARDS (acute respiratory distress syndrome) by reducing the host's inflammatory responses. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2820
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.