Learn More
Bioworld Gelzanâ„¢ (Gelrite), 1 kg
Supplier:  Bioworld 306340033
Encompass_Preferred
Gelzan™Synonym: GelriteAgar substitute produced from a bacterial substrate composed of glucaronic acid, rhamnose and glucose Aids in the detection of microbial contaminants Gel Strength: 400–700 g/cm2Recommended concentration: 3.0–5.0 g/L Gelling Temperature: 29–31°C Loss on drying: 6–16%Adjust Gelzan to the medium by means of a sieve to avoid clumpingGelzan™ is a trademark of the Monsanto Company
Catalog No. NC0802927
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.