Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GEM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100661
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100661 Supplier Invitrogen™ Supplier No. PA5100661
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GEM.

GEM belongs to the RAD/GEM family of GTP-binding proteins. It is associated with the inner face of the plasma membrane and could play a role as a regulatory protein in receptor-mediated signal transduction.The protein encoded by this gene belongs to the RAD/GEM family of GTP-binding proteins. It is associated with the inner face of the plasma membrane and could play a role as a regulatory protein in receptor-mediated signal transduction. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GEM
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GEM
Gene Accession No. P55040, P55041
Gene Alias AV020497; Gem; GTP binding protein (gene overexpressed in skeletal muscle); GTP binding protein overexpressed in skeletal muscle; GTP-binding mitogen-induced T-cell protein; GTP-binding protein GEM; GTP-binding protein overexpressed in skeletal muscle; kinase-inducible Ras-like protein; KIR; MGC26294; RAS-like protein KIR
Gene Symbols GEM
Host Species Rabbit
Immunogen A synthesized peptide derived from human GEM(Accession P55040), corresponding to amino acid residues V33-E83.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14579, 2669
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.