Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuTec Blades, Inc. GEM2 Facet Single Edge Blade Cartridge

Catalog No. 17989022
Change view
Click to view available options
Description:
Degreased
Material:
Stainless Steel, Plastic
No. per Case:
10 Cartridges/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Material No. per Case
17-989-022 Degreased Stainless Steel, Plastic 10 Cartridges/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-989-022 Supplier AccuTec Blades, Inc. Supplier No. 9404990000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Facet Single Edge Blade Cartridge is for use with Gem Runner™ Reloadable Safety Blade Dispenser in cutting of medical devices such as catheters.

Features:

  • Clean room ready clamshell packaging
  • Less oppourtunity for foreign material transfer or contamination from blade to finished product
  • Cartridge inserts into reloadable safety dispenser
  • Holds 100 blades

Specifications

Product Type 2 Facet
Material Stainless Steel, Plastic
Color Black
Description Degreased
Dimensions (L x W) 5.75 x 2.5 in.
For Use With (Application) For use with Gem Runner™ Reloadable Safety Blade Dispenser in cutting of medical devices such as catheters.
No. per Case 10 Cartridges/Cs.
No. per Pack 100 Blades/Cartridge
Weight (English) 0.68 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.