Learn More
Gemini Bio Products Gemcell Plus Human Serum, AB Serotype, Xeno-Free, 100 mL
SDP
Supplier:  Gemini Bio Products 100912100
Encompass_Preferred
GEMCELL PLUS U.S. Origin Human Serum is a premium xeno-free human serum designed for advanced therapy applications. It offers a rigorous non-reactive testing protocol and is manufactured from healthy male donors in FDA-licensed facilities. This ensures its high quality and safety for scientific research. Its unique formulation includes therapeutic grade human thrombin, enhancing cell culture workflows with consistent performance. Ideal for researchers seeking reliable and quality human serum.
Catalog No. NC2137067
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.