Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Gemfibrozil, 98%

Catalog No. AC461790050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461790050 5 g
AC461790250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461790050 Supplier Thermo Scientific Chemicals Supplier No. 461790050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 25812-30-0 | C15H22O3 | 250.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25812-30-0
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number MFCD00079335
InChI Key HEMJJKBWTPKOJG-UHFFFAOYSA-N
Synonym gemfibrozil, lopid, 5-2,5-dimethylphenoxy-2,2-dimethylpentanoic acid, jezil, decrelip, lipur, bolutol, apo-gemfibrozil, gemfibromax, cholespid
PubChem CID 3463
ChEBI CHEBI:5296
IUPAC Name 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid
SMILES CC1=CC=C(C)C(OCCCC(C)(C)C(O)=O)=C1

Specifications

Melting Point 60.0°C to 61.0°C
Quantity 5 g
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 250.33
Percent Purity 98%
Chemical Name or Material Gemfibrozil
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 247-280-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.