Description
- Unique design allows users to run up to 8 independently operated stations directly from a laptop using a standard USB.
- By allowing users to define a complete study in advance, Gemini eliminates run time data entry that could affect the accuracy of results.
- Scrambled shock provides more accurate responses
- Solid state feedback-controlled, constant current shocker uses 8 unique outputs to scramble the shock across the grid floor.
- Scrambling the shock prevents the animal from determining a pattern and leads to more accurate responses.
- Kinder, gentler, unconditioned stimulus - offers an alternative air puff unconditioned stimulus
- Optional Components
- Air puff stimulation unit (AIRSTIM)
- Additional test stations
- Mouse passive avoidance start box
- Grid floors (for rats or mice)
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Animal | Mouse, Rat |
| Model | Shock, Air stim., Add'l Stations |