Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GemPure™ Plus Water for Injection (WFI) Quality Water (USP/EP/JP Grade), GeminiBio™
SDP

Catalog No. 11852117
Change view
Click to view available options
Quantity:
20 L
4 x 4 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-852-117 4 x 4 L
11-852-128 20 L
11-852-116 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
For further manufacturing or research use only. Not for direct Therapeutic use.
Catalog No. 11-852-117 Supplier GeminiBio Supplier No. 9010024X4
Add to Cart
Add to Cart
For further manufacturing or research use only. Not for direct Therapeutic use.

Manufactured in an ISO 13485 and 21 CFR 820 compliant facility and available in multiple volumes.

GemPure™ Plus Water for Injection (WFI) Quality Water is designed to support the most demanding upstream and downstream bioprocess workflows from development through commercial production. Manufactured and tested to meet the monographs of the United States, European, and Japanese Pharmacopeias, this multicompendial grade simplifies global qualification strategies while supporting productivity and consistency.

  • Multi-compendial grade meeting USP, EP, and JP monograph specifications
  • Supports upstream and downstream bioprocessing across development and commercial manufacturing
  • Manufactured and tested for high purity, consistency, and reliability
  • Available in 4 L, 20 L, 200 L, and custom volumes

Suited for:

  • Global programs requiring USP, EP, and JP alignment
  • Clinical and commercial manufacturing workflows
  • Customers seeking to streamline qualification across regions

Chemical Identifiers

CAS 7732-18-5

Specifications

pH 5.0 to 7.0
Color Clear and Colorless
Conductivity <5.0 μS/cm
DNase DNase-free
Endotoxin <0.25 EU/mL
EU Ph Reference 169
Molecular Weight (g/mol) 18
ChEBI 15377
Percent Purity 0.99999
Grade USP
Concentration or Composition (by Analyte or Components) H2O
Sterility Sterile
Filtered Through 0.2 μM Sterilization Grade Filter
Density 1 g/mL
Certificate of Analysis Yes
Quantity 4 x 4 L
Purification Method Filtration RO/DI
Shipping Condition Ambient protect from freezing
Chemical Name or Material Water, Water For Injection Quality
Show More Show Less
ShelfLife 24 Months
EINECSNumber 231-791-2
Recommended Storage 2°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.