Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GemPure™ Select Purified Water, Sterile (USP Grade), GeminiBio™
SDP

Catalog No. 11852124
Change view
Click to view available options
Quantity:
20 L
6 x 1 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-852-124 200 L
11-852-114 6 x 1 L
11-852-113 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
For further manufacturing or research use only. Not for direct Therapeutic use.
Catalog No. 11-852-124 Supplier GeminiBio Supplier No. 901005203
Add to Cart
Add to Cart
For further manufacturing or research use only. Not for direct Therapeutic use.

Produced in an ISO 13485 and 21 CFR 820 compliant facility.

GemPure™ Select Purified Water, Sterile (USP Grade) is manufactured and tested in accordance with the USP monograph for Sterile Purified Water. This grade supports non-parenteral applications, non-active medical device manufacturing, and cleaning processes requiring sterile purified water.

  • Meets USP monograph requirements for Sterile Purified Water
  • Suitable for non-parenteral applications
  • Used in medical device manufacturing and cleaning processes
  • Available in 1L, 20L, 200L, and custom sizes

Chemical Identifiers

CAS 7732-18-5

Specifications

pH 5.0 to 7.0
Color Clear and Colorless
Conductivity <5.0 μS/cm
DNase DNase-free
Endotoxin <0.25 EU/mL
EU Ph Reference 169
Molecular Weight (g/mol) 18
ChEBI 15377
Percent Purity 0.99999
Grade USP
Concentration or Composition (by Analyte or Components) H2O
Sterility Sterile
Filtered Through 0.2 μM Sterilization Grade Filter
Density 1 g/mL
Certificate of Analysis Yes
Quantity 200 L
Purification Method Filtration RO/DI
Shipping Condition Ambient protect from freezing
Chemical Name or Material Water, Water For Injection Quality
Show More Show Less
ShelfLife 24 Months
EINECSNumber 231-791-2
Recommended Storage 2°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.