Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gemtuzumab Chimeric Recombinant Rabbit Monoclonal Antibody (hP67.6 (Gemtuzumab))
GREENER_CHOICE

Catalog No. PIMA547867
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547867 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547867 Supplier Invitrogen™ Supplier No. MA547867
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: Recognises human CD33.

Gemtuzumab ozogamicin is a recombinant humanized IgG4 kappa antibody which is conjugated with calicheamicin derivative, a cytotoxic antitumor antibiotic isolated from fermentation of Micromonospora echinospora ssp. calichensis. Gemtuzumab ozogamicin has approximately 50% of the antibody loaded with 4-6 moles calicheamicin per mole of antibodyLabel. The antibody is specifically directed against the CD33 antigen present on leukemic myeloblasts in most patients with acute myeloid leukemia (AML). By binding to the CD33 antigen on tumors, the cytotoxic agent blocks the growth of cancerous cells and causes cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gemtuzumab Chimeric
Applications Flow Cytometry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone hP67.6 (Gemtuzumab)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias CDP-771; CMA-676 (ADC); gemtuzumab ozogamicin; hP67.6; hP67.6-calicheamicin (ADC); WAY-CMA-676
Host Species Rabbit
Immunogen Gemtuzumab was generated by huminization of the murine P67.6 clone, raised against CD33-transfected FMY9S5 cells of human origin.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.