Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ GENAPOL™ X-100, Protein Grade™ Detergent, 10% Solution, Sterile-Filtered, Calbiochem™,

Catalog No. 34579850ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-579-850ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-579-850ML Supplier MilliporeSigma™ Supplier No. 34579850ML
Add to Cart
Add to Cart

Specifications

Color Colorless
Quantity 50 mL
Packaging Glass Bottle
Synonym Polyoxyethylene (10) Isotridecyl Ether
Grade Protein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.