Learn More
Genscript Corporation Antibody Enzyme Other Products Molecular Biology 1 MG 1/EA
Supplier:  Genscript Corporation Z035021
Encompass_Preferred
Recombinant Acidaminococcus sp. BV3L6 Cas12a (cpf1) nuclease is expressed in E. coli and purified. The nuclease contains nuclear localization sequence (NLS) at the C-terminus and 6× His-tag at the C-terminus.
Catalog No. 50-252-9624
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.