Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ GeneAmp™ 10X PCR Gold Buffer & MgCl2

Catalog No. 4379874
Encompass_Preferred
Change view
Click to view available options
Volume:
6 x 1.5 mL
2 x 75 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume
43-798-74 2 x 75 mL
43-068-98 6 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-798-74 Supplier Applied Biosystems™ Supplier No. 4379874
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Applied Biosystems GeneAmp 10X PCR Gold Buffer is formulated to provide flexible, efficient activation of AmpliTaq Gold DNA Polymerase, resulting in highly specific, robust PCR amplification.

Applied Biosystems GeneAmp 10X PCR Gold Buffer and MgCl2 is formulated to provide flexible, efficient activation of AmpliTaq Gold DNA Polymerase, resulting in highly specific, robust PCR amplification. The ionic strength and pH of GeneAmp 10X PCR Gold Buffer has been optimized to provide a wider activation temperature and time range when used in with AmpliTaq Gold DNA Polymerase.

When GeneAmp 10X PCR Gold Buffer and the MgCl2 solution are used together, there is greater versatility in optimizing magnesium ion concentrations for optimal PCR amplification with any specific set of primers and DNA template.

Features

  • Magnesium ion–free
  • Gelatin-free
  • High versatility in optimizing magnesium ion concentration
  • MgCl2 solution provided separately
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Content And Storage • 10X GeneAmp PCR Gold Buffer, 2 x 75 mL
• 25 mM MgCl2, 2 x 75 mL

Store at -20°C.
Format Kit
Polymerase AmpliTaq Gold DNA Polymerase
Product Type 10X PCR Gold Buffer
Quantity 2 x 75 mL
Shipping Condition Wet Ice
For Use With (Application) Standard PCR
Volume 2 x 75 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.