Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GeneArt™ CRISPR Nuclease Vector with OFP Reporter Kit (with competent cells)

Catalog No. A21178
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21178 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For research use only. Not for use in diagnostic procedures.
Catalog No. A21178 Supplier Invitrogen™ Supplier No. A21178
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For research use only. Not for use in diagnostic procedures.

Includes

  • CRISPR OFP Nuclease Vector, linearized
  • 10X Oligonucleotide Annealing Buffer
  • DNase/RNase-free Water
  • 5X Ligation Buffer
  • T4 DNA Ligase
  • U6 Forward Sequencing Primer
  • Control, double stranded oligonucleotide
  • OneShot(TM) TOP10 Chemically Competent Cells

Simple, ready-to-use, all-in-one expression vector system for CRISPR-based genome editing.

Simple, ready-to-use, all-in-one expression vector system for expression of the functional components needed for CRISPR-Cas9 genome editing in mammalian cells with an orange fluorescent protein (OFP) reporter.

GeneArt CRISPR Nuclease Vector with OFP Reporter Kit is a vector system for expression of the functional components needed for CRISPR-Cas9 genome editing in mammalian cells with an orange fluorescent protein (OFP) reporter. The OFP reporter allows for fluorescence-based tracking of transfection efficiency, as well as FACS-based sorting/enrichment of Cas9 and CRISPR expressing cells.

The linearized GeneArt™ CRISPR nuclease vectors provide a rapid and efficient way to clone double-stranded oligonucleotides encoding a crRNA representing a desired target into an expression cassette that allows sequence-specific targeting of the Cas9 nuclease. Competent cells included for added convenience and highest cloning efficiencies.

GeneArt™ CRISPR Nuclease Vector system offers:
  • Easy-to-design genome engineering system
  • Affordable, ready-to-use cloning vectors
  • Enrichment for hard to transfect or difficult to modify cell
For research use only. Not for use in diagnostic procedures.

Certifications

CoA

Compliance

CoA

Recommended Storage

Store in freezer (-5 to -30°C)

TRUSTED_SUSTAINABILITY

Specifications

Cloning Method Restriction Enzyme/MCS
Content And Storage • CRISPR OFP Nuclease Vector, linearized
• 10X Oligonucleotide Annealing Buffer
• DNase/RNase-free Water
• 5X Ligation Buffer
• T4 DNA Ligase
• U6 Forward Sequencing Primer
• Control, double stranded oligonucleotide
Store the above at -5 to -30°C.

• OneShot™ TOP10 Chemically Competent Cells
Store at -80°C.
Detection Method Primer-probe
Form Liquid
Format Kit
Reaction Speed Fast
Technique CRISPR-Cas9
Product Line GeneArt
Product Type Crispr Nuclease Vector With ofP Reporter Kit
Promoter U6, CMV
Protein Tag OFP fusion
Quantity 10 reactions
Reporter Gene OFP
Sufficient For 10 Reactions
For Use With (Application) Gene Expression, Cloning
No. of Reactions 10 Reactions
Overhang 3' overhangs
Starting Material Oligos (DNA)
Show More Show Less
What is CRISPR-STOP?

CRISPR-STOP is a method of inserting STOP codon sequences to generate knockouts.

Please refer to the following article: CRISPR-STOP: gene silencing through base-editing-induced nonsense mutations.

How do I check for off-target effects in my CRISPR-modified cell lines?

The only complete way to confirm that there are no off-target effects is to sequence the entire genome of your cell. Alternatively, a less thorough means of checking for off-target editing is to perform targeted sequencing of sequences with the highest probability of off-target effects (i.e., most similar to your CRISPR target region).

How many guide RNAs do you recommend designing against my desired edit locus?

A single guide RNA (gRNA) is all that is required for targeting, but we do recommend testing 2-3 gRNAs against each locus being targeted for cleavage. Testing multiple gRNAs increases the chances of finding a gRNA with high editing efficiency, which will reduce the screening time required to identify the clone of interest.

What transfection methods do you recommend when working with your CRISPR products?

For transfecting of the Cas9 protein, we would recommend using the Neon transfection system or Lipofectamine CRISPRMAX Cas9 Transfection Reagent. For transfection of mRNA, we would recommend using Lipofectamine MessengerMAX Transfection Reagent. For transfection of CRISPR vectors, we would recommend using Lipofectamine 3000 Transfection reagent.

Do you have CRISPR products optimized for plants?

Our CRISPR products are optimized for mammalian systems, and have not been optimized for plant systems. However, we know researchers are doing that kind of work. While we have not tested our CRISPR products for plants, our new protein format would be ideal since it does not need to be translated and transcribed in the cell, so no plant-specific promoters required.

What are the benefits of using TAL/TALEN-based genome editing compared to your CRISPR system?

Invitrogen GeneArt Precision TALs, in addition to gene deletion, down-regulation and integration, can also be used for gene activation. Additionally, the system is based on a protein-DNA system, in contrast to CRISPR, which is based on a RNA-DNA system. TALs can be used to target any gene in any cell, including mammalian, bacterial, yeast, plants, insect, stem cells and zebrafish. Lastly, off-target effects are low when using the TAL system. Please refer to the following paper (http://www.sciencedirect.com/science/article/pii/S016816561500200X) where the authors compared TALs and CRISPR technology.

I am using the Invitrogen GeneArt CRISPR Nuclease Vector Kit and am getting very few ampicillin-resistant colonies on my selection plate. What could be the cause of this? Do you have any suggestions?

Here are possible reasons and solutions:

- Single-stranded (ss) oligonucleotides designed incorrectly; Make sure that each ss oligonucleotide contains the 5 nucleotides on the 3′ end required for cloning into the Invitrogen GeneArt CRISPR Nuclease Vector: ie., Top strand includes GTTTT on the 3′ end; Bottom strand includes CGGTG on the 3′ end.

- Double-stranded (ds) oligonucleotides were degraded; Store the 5 nM ds oligonucleotide stock in 1X Oligonucleotide Annealing Buffer. Avoid repeated freeze/thaw cycles; aliquot the 5 nM ds oligonucleotide stock and store at -20°C.

- Oligonucleotide annealing reaction inefficient; Ensure that the annealing reaction was performed as directed. If ambient temperature is >25°C, incubate the annealing reaction in a 25°C incubator.

I am using the Invitrogen GeneArt CRISPR Nuclease Vector Kit. I am having problems sequencing my construct to verify that my ds oligonucleotide insert is present and has the correct sequence. What do you recommend?

Please see the following recommendations:

- Use high-quality, purified plasmid DNA for sequencing. We recommend preparing DNA using the Invitrogen PureLink HQ Mini Plasmid Purification Kit. Add DMSO to the sequencing reaction to a final concentration of 5%.
- Increase the amount of template used in the sequencing reaction (up to twice the normal concentration).
- Use a 7:1 molar ratio of dITP:dGTP in your sequencing reaction.

How can I increase my efficiency of genome editing using the CRISPR-Cas9 technology?

There are several ways to increase efficiency, for instance, adding antibiotic selection and/or FAC sorting to enrich for the transfected cells will both help.

I am interested in using the CRISPR-Cas9 technology for genome editing. However, there is no 5’ NGG (PAM) sequence close to my locus of interest. What can I do?

PAM is a necessary requirement for CRISPR gene editing. However, in its absence, we recommend engineering a TAL effector to edit your desired gene efficiently. We offer GeneArt PerfectMatch TAL effectors. These are TAL effector nucleases that remove the 5´ base constraint and can be designed to target any desired sequence within the genome. Please go here for further details: https://www.thermofisher.com/us/en/home/life-science/genome-editing/geneart-tals.html

I am interested in the CRISPR-Cas9 technology but am worried about the nonspecific activity of Cas9. Are there best practices for avoiding off-target effects?

Carefully designed crRNA target oligos and avoiding homology with other regions in the genome are critical for minimizing off-target effects.

Can I perform multiplexed transfection of CRISPR gRNAs using the Invitrogen Neon Transfection System?

Yes, the Neon system does work for multiple gRNAs transfected at the same time.

What is the suggested ratio of Cas9 mRNA to IVT gRNA?

We recommend starting at a ratio of 0.5 µg of Cas9 mRNA and 50 ng of each IVT gRNA per well in a 24-well format. You should determine the optimal ratio for your particular cell line via a dose-response study.

I am interested in multiplexing. How can I do this using the GeneArt CRISPR system?

Create multiple gRNAs targeting the targets of your choice, followed by co-transfection with GeneArt CRISPR Nuclease mRNA or GeneArt Platinum Cas9 Nuclease. To make the gRNAs for Cas9 mRNA, use GeneArt CRISPR Strings DNA, U6 or IVT gRNAs (generated using either GeneArt CRISPR Strings DNA, T7 or the GeneArt Precision gRNA Synthesis Kit). For the Cas9 protein, use IVT gRNAs (generated using either GeneArt CRISPR Strings DNA, T7 or the GeneArt Precision gRNA Synthesis Kit).

Is it possible to insert a selection marker, e.g., neomycin or a fluorescent marker, into a preexisting CRISPR-Cas9 plasmid?

Yes, if you use the current Invitrogen GeneArt CRISPR nuclease vectors the respective Limited-Use Label Licenses (LULLs) will apply.

I see other publications using different PAM sequences such as NNNGATT, NNAGAA, or NAAAAC. Can I use that instead of NGG?

No, the PAM sequence is unique to the bacterial species that was used to create the Cas9. In the Invitrogen GeneArt kits, we derived Cas9 from Streptococcus pyogenes.

I am using the CRISPR-Cas9 technology. What is the PAM sequence?

PAM stands for the protospacer adjacent motif. It is necessary for Cas9 to bind to the DNA successfully. The PAM sequence for the Streptococcus pyogenes Cas9 in the Invitrogen GeneArt CRISPR kits is NGG.

I am interested in using the CRISPR system to modify my gene of interest using HDR instead of NHEJ repair. How can I do this?

With the CRISPR-Cas9 editing complex (DNA vector, mRNA or Protein), co-transfect a DNA repair template that contains high homology to the sequence of interest along with the desired sequence you would like to introduce into the DNA. By doing so HDR can occur, and your specific edits (mutation, insertion, etc.) can be incorporated into the genome.

How can I tell if my CRISPR genome editing experiment is working?

Cleavage efficiency can be detected using the Invitrogen GeneArt Genomic Cleavage Detection Assay. This assay relies on mismatch detection endonucleases to detect insertions and deletions (indels) generated during cellular NHEJ repair.

What is the excitation and emission of OFP? What laser and emission filters should I use for detection?

OFP has a peak excitation of 548 nm, and emission of 560 nm. We recommend a 488 nm laser for efficient excitation. Standard 530/30, 647/26, and 603/48 emission filters are recommended for detection.

What promoter is used to drive gRNA expression in the Invitrogen GeneArt CRISPR vectors?

gRNA expression is driven by the Pol III U6 promoter.

For how long are the reagents in the Invitrogen GeneArt CRISPR Nuclease Vector kit guaranteed?

All reagents are guaranteed to be stable for 6 months when properly stored. The vector, buffers, control oligo, plasmid, water, and sequencing primers should be stored at -20°C.

What reagent do you recommend for transfection of the CRISPR plasmids for cell culture?

The method of transfection varies based on cell type. For high-efficiency transfection in a broad range of mammalian cell lines, we recommend using the cationic lipid-based Invitrogen Lipofectamine 3000 Reagent.

What should I use to purify my CRISPR plasmid before transfection into eukaryotic cells?

We recommend using high-quality DNA prepared with the Invitrogen PureLink HiPure Plasmid Miniprep Kit. Ensure that DNA is free of contamination with phenol and sodium chloride.

After ligation and transformation of my CRISPR plasmid, how should I analyze my transformants?

Confirm the identity of the ds oligonucleotide insert in positive transformants by sequencing. Analyze each CRISPR nuclease construct to verify:

- That the ds oligonucleotide insert is present, and in the correct orientation
- That the ds oligonucleotide insert has the correct sequence

Note: Restriction analysis is not recommended due to the small size of the ds oligonucleotide insert.

Can I use my own competent cells to transform my Invitrogen GeneArt CRISPR nuclease vector after ligation?

Yes, we provide kits both with or without competent cells. However, the vectors were optimized using Invitrogen One Shot TOP10 chemically competent E. coli, designed for high-efficiency cloning and plasmid propagation and stable replication of high-copy number plasmids. Please note that using competent cells of different genotype may lower cloning efficiency and can also result in a higher proportion of vectors without insert

How do I design and order the CRISPR oligos for cloning into the nuclease vectors?

Start off by designing two single-stranded DNA oligonucleotides (24-25 bp), one encoding the target-specific crRNA (forward-strand oligonucleotide) and the other its complement (reverse complement-strand oligonucleotide). You can generate a double-stranded oligonucleotide suitable for cloning into the linearized vector provided in the kit by simply annealing the complementary oligonucleotides. Here are some guidelines:

- Length: Choose a target sequence ranging from 19 to 20 nucleotides in length that is adjacent to an NGG proto-spacer adjacent motif (PAM) sequence on the 3′ end of the target sequence. The 5′ G required for transcription initiation from the U6 Pol III promoter is already included in the vector overhangs and does not need to be included in the target sequence. Note: Do not include the PAM sequence in the oligonucelotide primers.
- Homology: Make sure that the target sequence does not contain significant homology to other genes, as this can increase off-target effects. Recently published work has shown that gRNA-Cas9 complexes can potentially tolerate 1-3 or more mismatches, depending on their location in the gRNA. Refer to published articles for more insights into choosing a target sequence (Fu et al., 2013; Mali et al., 2013).
- Orientation: You may choose a target sequence encoding either the sense sequence of the target locus or the antisense sequence. Thus, you can generate CRISPR RNA in two possible orientations, provided that it meets the PAM requirements on the 3′ end.

What are the benefits of using a CD4 reporter in the Invitrogen GeneArt CRISPR Nuclease Vector with CD4 Reporter Kit?

Populations of cells transfected with Invitrogen GeneArt CRISPR nuclease vectors containing CD4 reporters may be enriched using FACs or using Invitrogen Dynabeads CD4 magnetic beads.

What are the benefits of using an OFP reporter in the Invitrogen GeneArt CRISPR Nuclease Vector with OFP Reporter Kit?

The OFP reporter allows for fluorescence-based tracking of transfection efficiency, as well as FACS-based sorting/enrichment of Cas9- and CRISPR-expressing cells.

What promoter is driving expression of Cas9?

Our vectors contain a CMV promoter to drive expression of the Cas9 endonuclease.

When I receive my CRISPR oligonucleotide, what should I resuspend it in?

Either nuclease-free water or TE buffer is fine, but please anneal using the appropriate Oligonucleotide Annealing Buffer for your annealing reaction. Subsequently, please serially dilute your primers in a final concentration of 1X Oligonucleotide Annealing Buffer.

When designing my primers for the CRISPR oligonucleotide, what level or purity is recommended? And do the oligonucleotides need to be phosphorylated?

5′-nonphosphorylated desalted oligonucleotides are fine. However, HPLC or higher purity will increase the cloning efficiency of your double-stranded oligonucleotide into the Invitrogen GeneArt CRISPR Nuclease vector.

What are the advantages of using the Invitrogen GeneArt CRISPR Nuclease Vectors?

This system allows you to edit and engineer the genomic locus of your choice in a sequence-specific manner from a single plasmid. We offer two plasmids that can be used with these kits, with either the OFP reporter or CD4 ereporter, for ease of selection and enrichment by either FACs or Invitrogen Dynabeads technology. Both kits include all-in-one vectors that co-express both the noncoding guide RNA (including crRNA and tracrRNA) and the Cas9 endonuclease.

When should I use the Invitrogen GeneArt CRISPR Nuclease Vector system as opposed to the Invitrogen GeneArt CRISPR Nuclease mRNA system?

We recommend using the Invitrogen GeneArt CRISPR Nuclease Vector system when working with mammalian cells and in scenarios where there are no promoter constraints. We recommend using the Invitrogen GeneArt CRISPR Nuclease mRNA system when working with difficult-to-transfect cell lines, microinjections and for multiplexing.

What is an indel?

An indel refers to the genomic insertion or deletion of bases, which are incorporated during either cellular NHEJ or HD repair mechanisms.

Can you elaborate on your suggestion to use CRISPR as a screening tool before going to TALs?

Since cleavage efficiency at a particular locus depends on the accessibility of the locus, chromatin state, and sequence, it is advisable to test multiple different loci/regions within a gene of interest. With CRISPR-Cas9-mediated genome editing, for each target of interest the user needs only to change the 19-20 bp target-specific oligo. After the cell lines have been screened and the sequence/locus with the highest cleavage efficiency has been identified, the biologically relevant mutations can be precisely created with high-specificity Invitrogen GeneArt TALs (https://www.thermofisher.com/us/en/home/life-science/genome-editing/geneart-tals.html).

I am using the CRISPR-Cas9 technology. How precise is the cleavage event?

Cleavage is precise, and, after binding of the Cas9 and gRNA complex to the target genomic sequence, the nuclease activity occurs 3 bases upstream of the PAM (NGG) site.

I am using the CRISPR technology to knock out a gene. How long after transfection should I assess mRNA levels and protein knockdown?

This would depend upon the half-life of the particular transcript in your cell. We typically start seeing reduction in mRNA levels as early as 24 hrs post transfection, with further reduction after 48-72 hrs. Hence, we recommend performing the genomic cleavage detection assay 48-72 hours post transfection.

What happens to Cas9 after it performs its endonuclease activity?

Cas9 is transiently expressed and will therefore disappear over time with successive cell divisions.

Is it possible to use CRISPR-Cas technology for prokaryotic gene engineering?

Yes, it is possible but our system is not for prokaryotes, and has only been optimized for mammalian systems. Please also consult our CRISPR custom services for further inquiries (custom.services@lifetech.com).

Have you tested the CRISPR system in hosts other than mammalian systems?

We have only tested these in mammalian systems (human and mouse cells).

Do you offer a service to generate custom cell lines using CRISPR?

Yes, we do offer this service (https://www.thermofisher.com/us/en/home/life-science/genome-editing/genome-engineering-services/cell-line-engineering-services.html).p>

I am using the CRISPR technology. I would like to design a guide RNA (gRNA) to insert a selection cassette such as neomycin. How can I do this?

The gRNA oligo design strategy in the Invitrogen GeneArt CRISPR Nuclease User Guide (https://tools.thermofisher.com/content/sfs/manuals/GeneArt_CRISPR_nuclease_mRNA_man.pdf)describes how you can design the guide RNA to target the locus in which the neomycin cassette should be inserted. The cassette (neomycin) can be inserted via HDR, in which case the neomycin cassette should contain locus specific homology arms.

I am trying to target a big and multifunctional protein using CRISPR technology. To what site should I design my sgRNA? How can I check that the editing occurred?

The first few exons would be best (closer to the promoter, resulting in premature transcript termination). Since the gRNA efficiency depends on the accessibility of the locus as well as the chromatin structure at that location, it is advisable to design and test a few target sites. Non-CRISPR-related mutations may be identified using gDNA isolated from non-CRISPR-treated cells as a control and performing a Invitrogen GeneArt Genomic Cleavage Detection Assay (https://www.thermofisher.com/order/catalog/product/A24372). Standard western blot analysis is a good measure for the verification of protein levels.

Can I use CRISPR technology to introduce large sequences, such as GFP or IRES?

Yes, this should be possible using CRISPR technology combined with HDR.

I am using the CRISPR technology and am worried about off-target effects. What is the best way to combat this issue?

Carefully designed crRNA target oligos and avoiding homology with other regions in the genome are critical for minimizing off-target effects.

What is the efficiency of HDR (homology directed repair) after the double-stranded break?

HDR efficiency is very low, on average less than 2%.

Can you provide suggestions on how to introduce a fragment into the genome or important sequence by HDR (homology directed repair)?

Create a double-stranded DNA break using the GeneArt CRISPR Nuclease Vector (https://www.thermofisher.com/us/en/home/life-science/genome-editing/geneart-crispr/crispr-nuclease-vector.html), while simultaneously transfecting your plasmid-based donor repair template. Your donor repair template plasmid will contain the sequence you wish to introduce that is flanked by at least 500 bp (or more) of sequence, which results in efficient homologous recombination of your sequence.

What is most efficient for HDR (homology directed repair), and what length is recommended for the homologous exogenous DNA, single-stranded oligos, double-stranded oligos, or large homologous arms (>1 kb) from a plasmid?

All of them may work, but for better efficiency, a longer homology arm is better (at least 500 bp (or more) on either side of the exogenous DNA). The homology length is dependent on the size of the fragment and will need to be tested. ssDNA may be error-prone or choose NHEJ. We offer the Invitrogen GeneArt Strings dsDNA fragments (1-3 kb) to assist with this type of application.

What is the difference between NHEJ- and HDR-mediated repair?

Both HDR (homology directed repair) and NHEJ (non-homologous end joining) are cellular mechanisms through which double-stranded DNA lesions are repaired. When a repair template is not present, NHEJ occurs to ligate double-stranded breaks, leaving behind insertion/deletion (indel) mutations. HDR is an alternative repair pathway in which a repair template is used to copy the sequence to the double-stranded break. You can, therefore, introduce specific nucleotide changes or DNA fragments into your target gene by using HDR with a repair template.

Are the indel events that happen in a particular locus identical between the cells? Can we include a clonal step before expanding and enriching the population?

Clonal isolation and a combined cleavage analysis and sequence verification of the edited clone is advisable.

What comprises the CRISPR-Cas system?

There are 3 components:

- The Cas nuclease Cas9 (a double-stranded DNA endonuclease)
- A target-complementary crRNA
- An auxiliary tracrRNA

The crRNA and the tracrRNA of the Invitrogen GeneArt CRISPR Nuclease Vector are expressed together as a guide RNA that mimics the natural crRNA-tracrRNA chimera in bacterial systems.

How does the Invitrogen GeneArt CRISPR-Cas system work?

As a simple two-component system that includes the Cas9 endonuclease and a noncoding guide RNA (gRNA), the engineered Type II CRISPR/Cas system can be leveraged to cleave genomic DNA at a predefined target sequence of interest. The gRNA has two molecular components: a target-complementary CRISPR RNA (crRNA) and an auxiliary trans-activating crRNA (tracrRNA). Both the gRNA and the PAM (NGG) motif guide the Cas9 nuclease to a specific genomic sequence to form a complex, followed by local strand separation (R-loop), at which the Cas9 nuclease creates a double-stranded DNA break (DSB) 3 nucleotides upstream from the PAM site. As a result, you may bring new functionality to the gene of interest via mutations, create knockouts, or introduce nonnative or synthetic genomic sequences to investigate novel applications.

CRISPR also allows for non-editing application flexibility such as gene regulation or RNAi-related studies. The Cas9 nuclease may be tethered to different functional domains (activators or repressors) or the gRNA may be designed to directly cleave miRNA.

What is the advantage of generating gene knockouts by TAL or CRISPR compared to vector-based stably expressed shRNA?

TAL and CRISPR directly edit the genome by a combined cleavage and repair mechanism to impart permanent genomic change (deletion or frameshift mutation), and the resulting gene knockouts are very efficient. RNAi technology, on the other hand, is an indirect method in either down-regulating or shutting down a gene completely through direct interaction with RNA (coding or noncoding). Even in the case for stably expressed miRNA or shRNA systems, it may be difficult to effect complete penetrance (i.e., shRNA:mRNA ratio) since knock-down levels are dependent on the activity of the promoter (related to integration location).

What is CRISPR technology used for?

With their highly flexible but specific targeting, CRISPR-Cas systems can be manipulated and redirected to become powerful tools for genome editing. CRISPR-Cas technology permits targeted gene cleavage and gene editing in a variety of eukaryotic cells, and because the endonuclease cleavage specificity in CRISPR-Cas systems is guided by RNA sequences, editing can be directed to virtually any genomic locus by engineering the guide RNA sequence and delivering it along with the Cas endonuclease to your target cell.

What does CRISPR and CRISPR-Cas stand for?

CRISPR stands for clustered regularly interspaced short palindromic repeat; CRISPR-Cas (CRISPR-associated) systems are used for genome editing in various host organisms.


WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.