Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GeneBLAzer™ ADORA2A CHO-K1 DA Assay Kit

Catalog No. K1345
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1345 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1345 Supplier Invitrogen™ Supplier No. K1345

missing translation for 'validMainframeMsg'

GeneBLAzer™ ADORA2A CHO-K1 DA Assay Kit

This cell line is available as dividing cells by requesting a quote for K1245 from dividingcells@invitrogen.com

GeneBLAzer™ ADORA2A CHO-K1 DA (Division Arrested) cells and ADORA2A-CRE-bla CHO-K1 cells contain the human Adenosine A2a (ADORA2A) receptor stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. no.K1129) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE). Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both ADORA2A CHO-K1 DA cells and ADORA2A-CRE-bla CHO-K1 cells are functionally validated for Z’-factor and EC50 concentrations of 5’-adenosine (NECA); (Figure 1). In addition, ADORA2A-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

The Adenosine A2a Receptor (ADORA2A) has been shown to have many roles throughout the body. ADORA2A is involved in pain and inflammation regulation, inhibition of platelet aggregation, blood pressure regulation and contributes to ischemic brain damage. Selective antagonists of the ADORA2A pathway are being tested as adjuvants to dopaminergic drugs to treat Parkinson’s disease and schizophrenia.


Storage:

GeneBLAzer™ ADORA2A CHO-K1 DA Cells and ADORA2A-CRE-bla CHO-K1 Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use.


Contents:
GeneBLAzer™ ADORA2A CHO-K1 DA Assay Kit
Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol).
Includes:
  • ADORA2A CHO-K1 DA cells
  • LiveBLAzer™-FRET B/G Loading Kit, 70μg
  • Solution D, 1ml
  • 2 protocols
  • Certificate of Analysis
    GeneBLAzer™ ADORA2A CHO-K1 DA cells
    Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
    Includes:
  • ADORA2A CHO-K1 DA cells
  • 1 protocol
  • Certificate of Analysis
    GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells
    (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
    Includes:
  • ADORA2A-CRE-bla CHO-K1 cells
  • 1 protocol
  • Certificate of Analysis
  • TRUSTED_SUSTAINABILITY

    Specifications

    Accession Number NM_000675
    Assay Entry Cell-based beta lactamase reporter gene
    Cell State Division-Arrested Cells
    Detection Method Fluorescence
    Gene ID (Entrez) 135
    Gene Symbol ADORA2A
    Population Status Single Cell Clone Origin
    Product Line GeneBLAzer
    Quantity 1 plate
    Reporter Gene BLA (Beta-Lactamase)
    Target Entry ADORA2A
    Type Division-Arrested Cells Assay Kit
    Cell Line CHO-K1
    System Type GeneBLAzer™
    Druggable Target GPCR
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.