Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ AVPR1A CHO-K1 DA Assay Kit

Catalog No. K1323
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1323 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1323 Supplier Thermo Scientific™ Supplier No. K1323

GeneBLAzer™ AVPR1A CHO-K1 DA (Division Arrested) cells and AVPR1A-NFAT-bla CHO-K1 cells contain the human Arginine Vasopressin 1a (AVPR1A) receptor stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.

This cell line is available as dividing cells by requesting a quote for Cat. No. K2186 from dividingcells@invitrogen.com

GeneBLAzer™ AVPR1A CHO-K1 DA (Division Arrested) cells and AVPR1A-NFAT-bla CHO-K1 cells contain the human Arginine Vasopressin 1a (AVPR1A) receptor stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. no.K1129) contain a beta-lactamase reporter gene under control of the nuclear factor of activated T-cells (NFAT) response element. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both AVPR1A CHO-K1 DA cells and AVPR1A-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of vasopressin, (Figure 1). In addition, AVPR1A-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

The family of receptors that recognize vasopressin and oxytocin consists of four members: vasopressin receptors 1a (AVPR1a), 1b (AVPR1b), and 2 (AVPR2), and the oxytocin (OT) receptor. AVPR1a, AVPR1b, and the OT receptor couple to the Gq/G11 class of G proteins and trigger the release of intracellular calcium when stimulated. AVPR2 receptors couple to Gs proteins and activate adenyl cyclase. All three AVPR receptors have high affinity for their endogenous ligand, vasopressin, but AVPR1b has less affinity for oxytocin than does AVPR1a or AVPR2. The hormones vasopressin and oxytocin are synthesized in the hypothalamus. Oxytocin causes contractions in the uterus during childbirth. The main functions of arginine vasopressin (AVP) are reabsorption of water by the kidneys and contraction of smooth muscle in arteries. This system may also play a role in memory and learning.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000706.3
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Detection Method Fluorescence
Gene ID (Entrez) 552
Gene Symbol AVPR1a
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 Plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry AVPR1A
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.