Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ AVPR2 CHO-K1 DA Assay Kit

Catalog No. K1329
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1329 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1329 Supplier Thermo Scientific™ Supplier No. K1329

GeneBLAzer™ AVPR2 CHO-K1 DA (Division Arrested) cells and AVPR2-CRE-bla CHO-K1 cells contain the human Arginine Vasopressin 2 (AVPR2) receptor stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.

CellSensor™ CRE-bla CHO-K1 cells (Cat. no.K1129) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE). Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate) or a tube of cells sufficient to analyze 10 x 384-well plates.

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both AVPR2 CHO-K1 DA cells and AVPR2-CRE-bla CHO-K1 cells are functionally validated for Z’-factor and EC50 concentrations of dDAVP (Figure 1). In addition, AVPR2-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

Agonists and antagonists of the AVPR2 receptors are in various stages of clinical trials for the treatment of hypertension and as diuretics.

Storage:

GeneBLAzer™ AVPR2 CHO-K1 DA Cells and AVPR2-CRE-bla CHO-K1 Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use.

Contents:
GeneBLAzer™ AVPR2 CHO-K1 DA Assay Kit
Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol).
Includes:
•AVPR2 CHO-K1 DA cells
•LiveBLAzer™-FRET B/G Loading Kit, 70μg
•Solution D, 1ml
•2 protocols
•Certificate of Analysis
GeneBLAzer™ AVPR2 CHO-K1 DA cells
Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
•AVPR2 CHO-K1 DA cells
•1 protocol
•Certificate of Analysis
GeneBLAzer™ AVPR2-CRE-bla CHO-K1 cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
•AVPR2-CRE-bla CHO-K1 cells
•1 protocol
•Certificate of Analysis

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000054
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Detection Method Fluorescence
Gene ID (Entrez) 554
Gene Symbol AVPR2
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry AVPR2
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.