Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ C5AR1-Gα15 CHO-K1 DA Assay Kit

Catalog No. K1544
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1544 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1544 Supplier Thermo Scientific™ Supplier No. K1544

The GeneBLAzer™ C5AR1-Gα15 CHO-K1 DA (Division Arrested) cells contain the human Complement Component 5a Receptor 1 (C5AR1) stably integrated into the GeneBLAzer™ Gα15-NFAT-bla CHO-K1 cell line.

The GeneBLAzer™ C5AR1-Gα15 CHO-K1 DA (Division Arrested) cells contain the human Complement Component 5a Receptor 1 (C5AR1) (Accession No. NM_000710) stably integrated into the GeneBLAzer™ Gα15-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gα15-NFAT-bla CHO-K1 (Cat. No. K1213) contains a beta-lactamase (bla) reporter gene under control of a NFAT response element and a promiscuous G Protein, Gα15, stably integrated into CHO-K1 cells.

The C5AR1-Gα15 CHO-K1 DA (Division Arrested) cells cells are functionally validated for Z'-factor and EC50 concentrations using C5a. In addition, C5AR1-Gα15-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_001736.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol).

Includes:
• GeneBLAzer™ C5AR1-Gα15 CHO-K1DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D
• GeneBLAzer™ C5AR1-Gα15 CHO-K1 Assay Protocol
• LiveBLAzer™-FRET B/G Loading Protocol
• Certificate of Analysis

The GeneBLAzer™ C5AR1-Gα15 CHO-K1 DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
Gene Symbol C5aR1
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 Plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry C5AR1
Type Division-Arrested Cells Assay Kit
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.