Learn More
Thermo Scientific™ GeneBLAzer™ CHO-K1 Control Kit
Description
The GeneBLAzer™ CHO-K1 Control Kit contains constitutively expressing (CMV-bla) CHO-K1 cells and wildtype CHO-K1 cells to provide positive and negative controls for evaluating beta-lactamase activity.
Specifications
Specifications
| Cell State | Dividing Cells |
| Content And Storage | The GeneBLAzer™ CHO-K1 Control Kit contains 2 x 1 mL vials of cells (∼3 x 106 cells/mL for CMV-bla CHO-K1 and ∼2 x 106 cells/mL for CHO-K1) and is shipped on dry ice. Store in liquid nitrogen upon receipt. |
| Detection Method | Fluorescence |
| Product Line | GeneBLAzer |
| Quantity | 1 Kit |
| Type | CHO-K1 Control Kit |
| Cell Line | CHO-K1 |
| System Type | CellSensor™ |
| Druggable Target | Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.