Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ CRHR1 CHO-K1 DA Assay Kit

Catalog No. K1367
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1367 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1367 Supplier Thermo Scientific™ Supplier No. K1367

GeneBLAzer™ CRHR1 CHO-K1 DA (Division Arrested) cells and CRHR1-CRE-bla CHO-K1 cells contain the human Corticotropin Releasing Factor 1 (CRHR1) receptor stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.

Division Arrested (DA) cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both CRHR1 CHO-K1 DA cells and CRHR1-CRE-bla CHO-K1 cells are functionally validated for Z’-factor and EC50 concentrations of corticotropin releasing factor (CRF), (Figure 1). In addition, CRHR1-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

Corticotropin releasing factor is a 41-amino acid peptide that plays a role in the integration of autonomic, neuroendocrine, and behavioral responses to stress. These effects are mediated through two receptor families, CRHR1 and CRHR2. While CRF was originally isolated from the hypothalamus, where it was shown to be the primary neuroregulator mediating the hypothalamic-pituitary-adrenocortical stress axis, it has since been found to be widely distributed outside the hypothalamus throughout the central nervous system. Presently, there are five distinct targets for CRF with unique pharmacology and localization. These have been placed into three distinct classes, two of which are the G-protein-coupled receptors CRF1 (CRHR1) and CRF2 (CRHR2). Three functional splice variants have been identified for the mammalian CRHR2 receptor, although pharmacological characterization of these splice variants has revealed no major differences between them.

GeneBLAzer™ CRHR1 CHO-K1 DA Cells and CRHR1-CRE-bla CHO-K1 Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use.

Contents:

GeneBLAzer™ CRHR1 CHO-K1 DA Assay Kit
Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol). Includes:
•CRHR1 CHO-K1 DA cells
•LiveBLAzer™-FRET B/G Loading Kit, 70μg
•Solution D, 1ml
•2 protocols
•Certificate of Analysis

GeneBLAzer™ CRHR1 CHO-K1 DA cells
Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
•CRHR1 CHO-K1 DA cells
•1 protocol
•Certificate of Analysis

GeneBLAzer™ CRHR1-CRE-bla CHO-K1 cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
Includes:
•CRHR1-CRE-bla CHO-K1 cells
•1 protocol
•Certificate of Analysis

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_004382.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Detection Method Fluorescence
Gene ID (Entrez) 1394
Gene Symbol CRHR1
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry CRHR1
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.