Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneBLAzer™ EDG3-Gα15 HEK 293T DA Assay Kit

Catalog No. K1319
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1319 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1319 Supplier Thermo Scientific™ Supplier No. K1319

GeneBLAzer™ EDG3-Gα15 HEK 293T DA (Division Arrested) cells and EDG3-Gα15-NFAT-bla HEK 293T cells contain the human EDG3 receptor stably integrated in the GeneBLAzer™ Gα15-NFAT-bla HEK 293T cell line.

This cell line is available as dividing cells by requesting a quote for K1234 from dividingcells@invitrogen.com

GeneBLAzer™ EDG3-Gα15 HEK 293T DA (Division Arrested) cells and EDG3-Gα15-NFAT-bla HEK 293T cells contain the human EDG3 receptor stably integrated in the GeneBLAzer™ Gα15-NFAT-bla HEK 293T cell line. GeneBLAzer™ Gα15-NFAT-bla HEK 293T cells (Cat. No. K1212) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element, and stably express the promiscuous G-protein, Gα15. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both EDG3-Gα15 HEK 293T DA cells and EDG3-Gα15-NFAT-bla HEK 293T cells are functionally validated for Z'-factor and EC50 concentrations of Sphingosine-1-phosphate (S1P), (Figure 1). In addition, EDG3-Gα15-NFAT-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

EDG-3(Endothelial-differentiation-gene-3)/S1P-3(Sphingosine-1-Phosphate-3) is a Gq/Gi/Go/G13/G12 coupled GPCR. EDG-3 has been shown to be responsible for bradycardia that has been induced in clinical trials as a side effect of the immunosuppressive drug FTY720 that targets EDG-1. EDG-3 has also been shown to induce vasodilation in response to sphingosylphosphorylcholine (SPC), sphingosine-1-phosphate (S1P), and lysosulfatide (LSF). The EDG-3 Receptor has also been shown to play a cooperative or redundant role in angiogenesis with EDG-1

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_005226.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage GeneBLAzer™ EDG3-Gα15 HEK 293T DA Assay Kit
Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol).
• EDG3-Gα15 HEK 293T DA cells (K1319A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D, 1 mL
• 2 protocols
• Certificate of Analysis

GeneBLAzer™ EDG3-Gα15 HEK 293T DA cells
Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• EDG3-Gα15 HEK 293T DA cells (K1320A)
• 1 protocol
• Certificate of Analysis

GeneBLAzer™ EDG3-Gα15-NFAT-bla HEK 293T cells
(LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol).
• EDG3-Gα15-NFAT-bla HEK 293T cells (K1234)
• 1 protocol
• Certificate of Analysis

Storage:
GeneBLAzer™ EDG3-Gα15 HEK 293T DA Cells and EDG3-Gα15-NFAT-bla HEK 293T Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use.
Detection Method Fluorescence
Gene ID (Entrez) 1903
Gene Symbol EDG3
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Target Entry EDG3
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.